Sodium stannate hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

14021-22 14021-36
CAAA14021-22EA 49.93 CAD
CAAA14021-22 CAAA14021-36
Sodium stannate hydrate, Reagent Grade
Sodium stannate hydrate

Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.


CAS No.: 12058-66-1 Molecular Formula: Na2SnO3.xH2O Formula Weight: 212.67(anhy) Physical Form: Crystalline Sensitivity: Air Sensitive Odor: Odorless Merck Reference: 14,8677 MDL No.: MFCD00014241

Formula: Na₂O₃Sn·H₂O
Melting Pt: 140 °C (dec.)
Density: 4.68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014241
CAS Number: 12058-66-1
EINECS: 235-030-5
Merck Index: 14,8677

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR