Potassium stannate trihydrate 95 Metals basis

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium tin(IV) oxide trihydrate

14015-30 14015-A1
CAAA14015-30EA 65.41 CAD
CAAA14015-30 CAAA14015-A1
Potassium stannate trihydrate 95 Metals basis
Potassium stannate trihydrate

Purity: 95+% CAS No.: 12125-03-0 Molecular Formula: K2SnO3.3H2O Formula Weight: 298.94 (244.89anhy) Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00150391

Formula: K₂O₃Sn·3H₂O
MW: 298.94 g/mol (244.89 anhy)
Melting Pt: 140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150391
CAS Number: 12125-03-0
EINECS: 235-255-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR