Cobalt disulfide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cobaltous sulphide, Cobalt (IV) sulphide, Cobalt (IV) sulfide, Cobalt(IV) sulphide, Cobalt(IV) sulfide, Cobaltous sulfide

14005-06
CAAA14005-06EA 276.5 CAD
CAAA14005-06
Cobalt disulfide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)
Cobalt disulphide

MDL: MFCD00064735


Purity: 99.5% CAS No.: 12013-10-4 Molecular Formula: CoS2 (may contain traces of other Co-S phases) Formula Weight: 123.06 Physical Form: -200 Mesh Powder Appearance: Black Odor: Odorless UN No.: 3077 MDL No.: MFCD00064735

Formula: CoS₂
MW: 123.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064735
CAS Number: 12013-10-4
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR