Arsenic, plasma standard solution, Specpure®, As 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13836-AD 13836-AE 13836-AP
CAAA13836-ADEA 83.74 CAD
CAAA13836-AD CAAA13836-AE CAAA13836-AP
Arsenic, plasma standard solution, Specpure®, As 1000µg/ml, Thermo Scientific
Standards Cationic Standards Arsenic Standards

Molecular Formula: As in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR