Ytterbium, Plasma Standard Solution, Specpure®, Yb 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13819-AE 13819-AP
CAAA13819-AEEA 129.88 CAD
CAAA13819-AE CAAA13819-AP
Ytterbium, Plasma Standard Solution, Specpure®, Yb 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Ytterbium Standards

MDL: MFCD00011286


Molecular Formula: Yb2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011286

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR