Sodium chloride, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

13683-06 13683-14
CAAA13683-06EA 180.12 CAD
CAAA13683-06 CAAA13683-14
Sodium chloride, ultra dry ≥99.998% (metals basis)
Sodium chloride

MDL: MFCD00003477 Fieser: 6,285


Purity: 99.998% CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.165 Refractive Index: 1.5442 Sensitivity: Hygroscopic Appearance: White Odor: Odorless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477

Formula: NaCl
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,8599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR