Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

13672-03 13672-06
CAAA13672-03EA 142.36 CAD
CAAA13672-03 CAAA13672-06
Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium (III) iodide

MDL: MFCD00049573 Fieser: 5,327 6,227


Purity: 99.99% CAS No.: 13813-40-6 Molecular Formula: TbI3 Formula Weight: 539.64 Physical Form: Flake, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049573

Formula: TbI₃
Melting Pt: 946 °C
Storage Temperature: Ambient
MDL Number: MFCD00049573
CAS Number: 13813-40-6
EINECS: 237-472-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR