Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304

Supplier: Thermo Scientific Chemicals

13478-CH 13478-RR 13478-RU
CAAA13478-CHEA 149.47 CAD
CAAA13478-CH CAAA13478-RR CAAA13478-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Stainless steel

Open area: 36%; Width of opening: 0.015in


Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Physical Form: Wire Cloth MDL No.: MFCD00148499

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR