Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 80 mesh woven from 0.127mm (0.005in) Type 304

Supplier: Thermo Scientific Chemicals

13477-CH 13477-RR 13477-RU
CAAA13477-CHEA 184.86 CAD
CAAA13477-CH CAAA13477-RR CAAA13477-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 80 mesh woven from 0.127mm (0.005in) Type 304
Stainless steel

Open area: 36%; Width of opening: 0.0075in


Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Physical Form: Wire Cloth MDL No.: MFCD00148499

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR