Calcium nitride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13124-06 13124-18 13124-30
CAAA13124-06EA 91.96 CAD
CAAA13124-06 CAAA13124-18 CAAA13124-30
Calcium nitride ≥99% (metals basis)
Calcium nitride

Purity: 99% CAS No.: 12013-82-0 Molecular Formula: Ca3N2 Formula Weight: 148.25 Physical Form: 12mm & Down Pieces Density: 2.63 Melting Point: 1195° UN No.: 3178 MDL No.: MFCD00015985

Formula: Ca₃N₂
Melting Pt: 1195 °C
Density: 2.63
Storage Temperature: Ambient
MDL Number: MFCD00015985
CAS Number: 12013-82-0
EINECS: 234-592-9
UN: 3178
ADR: 4.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR