Titanium(II) hydride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12857-06 12857-22 12857-36 12857-D3
CAAA12857-06EA 59.88 CAD
CAAA12857-06 CAAA12857-22 CAAA12857-36 CAAA12857-D3
Titanium(II) hydride ≥99% (metals basis)
Titanium (II) hydride

Powder, 325 mesh. Moisture sensitive. Purity based on metal contaminates only.


Purity: 99% CAS No.: 7704-98-5 Molecular Formula: H2Ti Formula Weight: 49.92 Physical Form: -325 Mesh Powder Density: 3.9 Appearance: Dark grey Odor: Odorless Melting Point: 450° dec. Merck Reference: 14,9473 UN No.: 1871 MDL No.: MFCD00151301

Formula: H₂Ti
Melting Pt: 450 °C (dec.)
Density: 3.9
Storage Temperature: Ambient
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,9473

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR