Tellurium ≥99.9999% (metals basis), broken ingot, Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

12607-14 12607-22 12607-36
CAAA12607-14EA 320.74 CAD
CAAA12607-14 CAAA12607-22 CAAA12607-36
Tellurium ≥99.9999% (metals basis), broken ingot, Puratronic®
Tellurium

MDL: MFCD00134062


Purity: 99.9999+% CAS No.: 13494-80-9 Odor: Odorless MDL No.: MFCD00134062

Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR