Tin 99,85 Metals basis, rods, Ø 6.35 cm (2.5 in)

Supplier: Thermo Scientific Chemicals

12601-36
CAAA12601-36EA 361.82 CAD
CAAA12601-36 CAAA12601-A3
Tin 99,85 Metals basis, rods, Ø 6.35 cm (2.5 in)
Tin

MDL: MFCD00133862


Purity: 99.85% CAS No.: 7440-31-5 Physical Form: ≈2.2cm/500g Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR