Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Danger

12397-FF 12397-FI
CAAA12397-FFEA 267.02 CAD
CAAA12397-FF CAAA12397-FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Gadolinium

MDL: MFCD00011022


Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈0.12g/25x25mm, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver UN No.: 3178 MDL No.: MFCD00011022

Formula: Gd
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR