Vanadium(III) oxide ≥95%

Supplier: Thermo Scientific Chemicals
Danger

12301-30 12301-A1 12301-A7
CAAA12301-30EA 148.36 CAD
CAAA12301-30 CAAA12301-A1 CAAA12301-A7
Vanadium(III) oxide ≥95%
Vanadium (III) oxide

MDL: MFCD00011455 Insoluble in water


Purity: 95% CAS No.: 1314-34-7 Molecular Formula: O3V2 Formula Weight: 149.88 Physical Form: Powder Density: 4.87 Appearance: Black Odor: Odorless Melting Point: 1970° Solubility: Insoluble in water Merck Reference: 14,9923 UN No.: 3285 MDL No.: MFCD00011455

Formula: O₃V₂
MW: 149.88
Melting Pt: 1970 °C
Density: 4.87
Storage Temperature: Ambient
MDL Number: MFCD00011455
CAS Number: 1314-34-7
EINECS: 215-230-9
UN: 3285
ADR: 6.1 II
Merck Index: 14,9923

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR