Zirconium nitride ≥99.5% (metals basis excluding Hf) (max. 3% Hf)

Supplier: Thermo Scientific Chemicals
Warning

12138-14 12138-22
CAAA12138-14EA 184.86 CAD
CAAA12138-14 CAAA12138-22
Zirconium nitride ≥99.5% (metals basis excluding Hf) (max. 3% Hf)
Zirconium nitride

MDL: MFCD00049642


Purity: 99.5% CAS No.: 25658-42-8 Molecular Formula: NZr Formula Weight: 105.23 Physical Form: -325 Mesh Powder Density: 7.09 Appearance: Yellow-brown Odor: Odorless Melting Point: 2980° UN No.: 3178 MDL No.: MFCD00049642

Formula: NZr
Melting Pt: 2980 °C
Density: 7.09
Storage Temperature: Ambient
MDL Number: MFCD00049642
CAS Number: 25658-42-8
EINECS: 247-166-2
UN: 3178
ADR: 4.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR