Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconium (IV) chloride

12104-09 12104-22 12104-36 12104-A3
CAAA12104-09EA 80.58 CAD
CAAA12104-09 CAAA12104-22 CAAA12104-36 CAAA12104-A3
Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade
Zirconium tetrachloride

Friedel-Crafts catalyst


Purity: 99.5+% CAS No.: 10026-11-6 Molecular Formula: ZrCl4 Formula Weight: 233.03 Physical Form: Powder Density: 2.803 Appearance: White to orange Melting Point: 331° subl. Solubility: Soluble in alcohol, ether. Decomposes in water forming ZrOCl2 and HCl Merck Reference: 14,10174 UN No.: 2503 MDL No.: MFCD00011306


Friedel-Crafts catalyst

Formula: Cl₄Zr
MW: 233.03 g/mol
Boiling Pt: 3577
Melting Pt: 331 °C (subl.)
Density: 2.803
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8 III
Merck Index: 14,10174

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR