Tungsten disulfide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Tungsten sulfide, Tungsten sulphide, Tungsten (IV) sulfide, Tungsten (IV) sulphide

11829-18 11829-30
CAAA11829-18EA 136.99 CAD
CAAA11829-18 CAAA11829-30
Tungsten disulfide ≥99.8% (metals basis)
Tungsten disulphide

MDL: MFCD00011467


Purity: 99.8% CAS No.: 12138-09-9 Molecular Formula: WS2 Formula Weight: 247.98 Physical Form: Powder Density: 7.5 Appearance: Black Melting Point: 1250° dec. MDL No.: MFCD00011467

Formula: S₂W
MW: 247.97
Melting Pt: 1250 °C (dec.)
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00011467
CAS Number: 12138-09-9
EINECS: 235-243-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR