Iron ≥99.995% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

11447-FF 11447-FI
CAAA11447-FFEA 442.4 CAD
CAAA11447-FF CAAA11447-FI
Iron ≥99.995% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Iron

MDL: MFCD00010999


Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈9.8g/25x25mm MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR