Aluminum phosphate ≥97%

Supplier: Thermo Scientific Chemicals
Danger

11109-30 11109-A1
CAAA11109-30EA 98.43 CAD
CAAA11109-30 CAAA11109-A1
Aluminum phosphate ≥97%
Aluminum phosphate

Applications: flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy
Insoluble in water and acetic acid; slightly soluble in concentrated hydrochloric acid and nitric acid


Purity: 97% CAS No.: 7784-30-7 Molecular Formula: AlO4P Formula Weight: 121.95 Physical Form: Powder Density: 2.566 Appearance: White Melting Point: 1500° Solubility: Insoluble in water and acetic acid; slightly soluble in concentrated hydrochloric acid and nitric acid UN No.: 3260 MDL No.: MFCD00003430


Applications: As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy

Formula: AlPO₄
Melting Pt: >1500 °C
Density: 2.566
Storage Temperature: Ambient
MDL Number: MFCD00003430
CAS Number: 7784-30-7
EINECS: 232-056-9
UN: 3260
ADR: 8,III
Merck Index: 14,358

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR