Silver nitrate ≥99.9995% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

10858-06 10858-14 10858-22
CAAA10858-06EA 84.53 CAD
CAAA10858-06 CAAA10858-14 CAAA10858-22
Silver nitrate ≥99.9995% (metals basis), Premion®
Silver nitrate

MDL: MFCD00003414


Purity: 99.9995% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odor: Odorless Melting Point: 212° Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414

Formula: AgNO₃
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 212 °C
Density: 4.352
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1,I
REACH: 01-2119513705-43
Merck Index: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR