Magnesium chloride hexahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

10797-14 10797-22
CAAA10797-14EA 317.58 CAD
CAAA10797-14 CAAA10797-22
Magnesium chloride hexahydrate ≥99.999% (metals basis), Puratronic®
Magnesium chloride hexahydrate

MDL: MFCD00149781 Fieser: 10,251


Purity: 99.999% CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Crystalline Aggregates Density: 1.570 Appearance: White Odor: Odorless Melting Point: 117° dec. Merck Reference: 14,5662 MDL No.: MFCD00149781

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C (dec.)
Density: 1.570
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR