Lithium iodide hydrate 99,995 Metals basis, Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium iodide hydrate

10738-06 10738-14
CAAA10738-06EA 103.02 CAD
CAAA10738-06 CAAA10738-14
Lithium iodide hydrate 99,995 Metals basis, Puratronic®
Lithium iodide hydrate

Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.


Purity: 99.995% CAS No.: 85017-80-7 Molecular Formula: LiI.xH2O Formula Weight: 133.84(anhy) Physical Form: Crystalline Aggregates Sensitivity: Light and Air Sensitive and Hygroscopic Merck Reference: 14,5535 MDL No.: MFCD00149769

Formula: ILi·H₂O
MW: 133.84 g/mol (anhy)
Melting Pt: 73 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149769
CAS Number: 17023-25-5
EINECS: 233-822-5
Merck Index: 14,5535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR