Lead(II) oxide ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead monoxide

10728-14 10728-22
CAAA10728-14EA 273.34 CAD
CAAA10728-14 CAAA10728-22
Lead(II) oxide ≥99.9995% (metals basis), Puratronic®
Lead (II) oxide

It is used in the manufacture of glass, ceramics and computer components.


Addition of lead oxide increases the refractive index, electrical resistivity, Curie temperature, and the ability of the glass to absorb X-rays. It is also added to decrease the viscosity of glass. It is employed in the vulcanization of rubber, lead acid batteries, and in the production of certain pigments and paints. In cathode ray tube glass lead oxide is used to block X-ray emission. It acts as an efficient reagent in oxidation reactions as well as condensation reactions of hydroxy ketones with diamines to form quinoxaline derivatives with high yield.

Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 888
Density: 9.53
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9 III
Merck Index: 14,5413

Order Now

Specification Test Results

Total Metal Impurities 0.0005% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR