Lead(II) acetate trihydrate ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10719-06 10719-18 10719-30
CAAA10719-06EA 34.6 CAD
CAAA10719-06 CAAA10719-18 CAAA10719-30
Lead(II) acetate trihydrate ≥99.995% (metals basis), Puratronic®
Lead (II) acetate trihydrate

MDL: MFCD00150023 Beilstein Registry No.: 3730298 Notes: Decomposes above 200°C


Purity: 99.995% CAS No.: 6080-56-4 Molecular Formula: Pb(OOCCH3)2.3H2O Formula Weight: 379.33 (325.28anhy) Physical Form: Crystalline Density: 2.55 Melting Point: 75° Beilstein Reference: 3730298 Merck Reference: 14,5397 UN No.: 1616 MDL No.: MFCD00150023

Formula: (H₃CCOO)₂Pb·3H₂O
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C
Density: 2.55
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 14,5397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR