Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10699-06 10699-14 10699-22 10699-36
CAAA10699-06EA 53.4 CAD
CAAA10699-06 CAAA10699-14 CAAA10699-22 CAAA10699-36
Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Copper (II) nitrate hydrate

MDL: MFCD00149669 Fieser: 1,163 2,87 12,231 15,101 18,115 19,110 20,121 21,148


Purity: 99.999% CAS No.: 13778-31-9 Molecular Formula: CuN2O6.xH2O Formula Weight: 187.55(anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Blue Odor: Odorless Merck Reference: 14,2643 UN No.: 3085 MDL No.: MFCD00149669

Formula: CuN₂O₆·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149669
CAS Number: 13778-31-9
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,2643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR