Bismuth(III) nitrate, hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10657-14 10657-22
CAAA10657-14EA 162.74 CAD
CAAA10657-14 CAAA10657-22
Bismuth(III) nitrate, hydrate ≥99.999% (metals basis), Puratronic®
Bismuth (III) nitrate

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.


Purity: 99.999% CAS No.: 10361-44-1 Molecular Formula: Bi(NO3)3.xH2O Formula Weight: 394.99(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Colorless to white Melting Point: 30° dec. Merck Reference: 14,1271 UN No.: 1477 MDL No.: MFCD00003460

Formula: Bi(NO₃)₃·5H₂O
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C (dec.)
Density: 2.736 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003460
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Merck Index: 14,1271

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR