Potassium tetracyanopalladate(II) hydrate

Supplier: Thermo Scientific Chemicals
Warning

10535-03
CAAA10535-03EA 323.9 CAD
CAAA10535-03
Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate

CAS No.: 14516-46-2 Molecular Formula: K2[Pd(CN)4].xH2O Formula Weight: 288.68(anhy) Physical Form: Crystalline Appearance: White MDL No.: MFCD00150431

Formula: K₂[Pd(CN)₄]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150431
CAS Number: 14516-46-2
EINECS: 238-520-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR