Diethyl vinylphosphonate 97%

Supplier: Thermo Scientific Chemicals
Danger

10368-09
CAAA10368-09EA 251.22 CAD
CAAA10368-09
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate

Purity: 97% CAS No.: 682-30-4 Molecular Formula: H2C=CHP(O)(OC2H5)2 Formula Weight: 164.14 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.068 Refractive Index: 1.4290 Boiling Point: 202° UN No.: 3278 MDL No.: MFCD00009079

Formula: C₆H₁₃O₃P
Boiling Pt: 202 °C
Density: 1.068
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009079
CAS Number: 682-30-4
EINECS: 211-663-2
UN: 3278
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR