Platinum ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

10283-FF 10283-FI 10283-FL 10283-GH
CAAA10283-FFEA 513.83 CAD
CAAA10283-FF CAAA10283-FI CAAA10283-FL CAAA10283-GH AA10283-MAT CAAA10283-MAT
Platinum ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)
Platinum

Platinum gauze, 52 mesh woven from 0.1 mm (0.004") dia. wire.


Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.47g/25x25mm, Wire Cloth Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR