Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

10228-FF
CAAA10228-FFEA 717.32 CAD
CAAA10228-FF
Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Lutetium

MDL: MFCD00011098


Purity: 99.9% CAS No.: 7439-94-3 Physical Form: ≈0.62g/25x25mm, Packaged in mylar under argon Appearance: Silver-grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00011098

Formula: Lu
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1651…1675 °C
Density: 9.849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
UN: 3178
ADR: 4.1,III
Merck Index: 12,05642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR