Cerium ≥99.9% (REO, rare earth oxide basis), foil, thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals
Danger

10139-FF 10139-FI
CAAA10139-FFEA 398.16 CAD
CAAA10139-FF CAAA10139-FI
Cerium ≥99.9% (REO, rare earth oxide basis), foil, thickness 1.0 mm (0.04 in)
Cerium

MDL: MFCD00010924


Purity: 99.9% CAS No.: 7440-45-1 Physical Form: ≈4.2g/25x25mm, Packaged in mylar under argon Appearance: Silver-grey Odor: Odorless UN No.: 1333 MDL No.: MFCD00010924

Formula: Ce
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 795 °C
Density: ∼6,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR