Aluminium ≥99.9% (metals basis), granules 8-12 mm (0.32-0.47 in)

Supplier: Thermo Scientific Chemicals

10093-30 10093-A1 10093-A7
CAAA10093-30EA 83.9 CAD
CAAA10093-30 CAAA10093-A1 CAAA10093-A7
Aluminium ≥99.9% (metals basis), granules 8-12 mm (0.32-0.47 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9% CAS No.: 7429-90-5 Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR