Allylpalladium(II) chloride dimer (≥ 56,0% Pd)

Supplier: Thermo Scientific Chemicals
Warning

10005-01 10005-03 10005-06
CAAA10005-01EA 95.59 CAD
CAAA10005-01 CAAA10005-03 CAAA10005-06
Allylpalladium(II) chloride dimer (≥ 56,0% Pd)
Allylpalladium(II) chloride dimer

CAS No.: 12012-95-2 Molecular Formula: C6H10Cl2Pd2 Formula Weight: 365.85 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow Melting Point: 120° dec. Solubility: Very soluble in dichloromethane and dichloroethane. Slightly soluble in toluene, insoluble in water Beilstein Reference: 4124623 MDL No.: MFCD00044874

Formula: C₆H₁₀Cl₂Pd₂
Melting Pt: 120 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00044874
CAS Number: 12012-95-2
EINECS: 234-579-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR