Tantalum ≥99.98% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

00933-KF 00933-KJ
CAAA00933-KFEA 71.1 CAD
CAAA00933-KF CAAA00933-KJ
Tantalum ≥99.98% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Tantalum

MDL: MFCD00011252


Purity: 99.98% CAS No.: 7440-25-7 Physical Form: ≈6.8g/slug Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR