PfuTurbo DNA Polymerase, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600254 600324 600414 600256 600602 600250 600252 600255 600320 600322 600410 600412 600600
CA97066-444EA 1236.52 CAD
CA97066-444 CA97066-450 CA99901-814 CA99902-324 CA99902-380 CAST600250 CAST600252 CAST600255 CAST600257 CAST600259 CAST600320 CAST600322 CAST600410 CAST600412 CAST600600
PfuTurbo DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
This DNA polymerase is ideal for a variety of techniques requiring high-fidelity DNA synthesis by the polymerase chain reaction (PCR).
PfuTurbo DNA Polymerase is an enhanced version of Pfu DNA Polymerase for robust, high-fidelity PCR. PfuTurbo DNA polymerase is a mixture of cloned Pfu DNA polymerase and the exclusive thermostable ArchaeMaxx polymerase-enhancing factor that enhances PCR product yields and increases target length capability without altering DNA replication fidelity.
PfuTurbo DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuTurbo DNA Polymerase for robust, high-fidelity PCR.
PfuTurbo Hotstart DNA Polymerase is a special formulation of high fidelity PfuTurbo DNA Polymerase with hot start activity.
PfuTurbo Cx Hotstart DNA Polymerase is a specially formulated with a mutant of Pfu DNA Polymerase designed to overcome uracil sensitivity for more robust performance.
The PfuTurbo Hotstart PCR Master Mix is a 2X formulation of PfuTurbo hotstart DNA Polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more