Recombinant Ribonuclease Inhibitor

Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA

300151 300152
CA99901-510EA 247.7 CAD
CA99901-510 CA99901-512
Recombinant Ribonuclease Inhibitor
Proteins and Peptides

Purified Ribonuclease Inhibitor tested to be free of contaminating activity for the degradation of nucleic acids.


  • Unit Definition: One unit will inhibit 50 % of the activity of 5 ng of RNase A


RNase Block Ribonuclease Inhibitor is a 50 kDa protein that specifically inhibits common eukaryotic RNases including RNase A, RNase B, and RNase C. It is a recombinant ribonuclease inhibitor derived by a molecular cloning technology that inactivates RNases by binding noncovalently to the RNase molecule. Only 25 to 40 U is required for a standard transcription reaction.


Caution: For research use only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR