HiScreen Q FF Columns
Supplier: Cytiva
28950510
CA97067-816EA 357.1 CAD
CA97067-816
HiScreen Q FF Columns
Chromatography Columns
HiScreen Q FF columns are prepacked with Q Sepharose Fast Flow, a strong anion exchanger, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.
  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
More Product Information