Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
10-013-LX 10-013-LB 15-013-CM 15-013-LX 15-013-LB 15-017-CV 15-017-CM 15-018-CV 15-018-CM 10-014-CV 10-014-CM 17-205-CV 17-204-CI 17-206-CI 50-003-PB 50-003-PC 50-013-PB 50-013-PC 90-013-PB 90-113-PB 17-207-CV 10-013-CV 10-013-CM 10-017-CV 10-017-CM 15-013-CV 10-090-CM 10-101-CV 10-102-CV 10-027-CV
CA45000-308EA 1013.14 CAD
CA45000-308 CA45000-310 CA45000-318 CA45000-320 CA45000-322 CA45000-324 CA45000-326 CA45000-328 CA45000-330 CA45000-332 CA45000-334 CA45000-336 CA45000-340 CA45000-342 CA45000-518 CA45000-520 CA45000-522 CA45000-524 CA45000-526 CA45000-534 CAMT-17-207-CV CA45000-304 CA45000-306 CA45000-312 CA45000-314 CA45000-316 CA97065-362 CA71002-872 CA71002-874 CA89500-458
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Cell Culture Media
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Low endotoxin - <0.25 EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
The original formulation was used to culture embryonic mouse cells and has since been modified to support a variety of normal and transformed cells, such as primary cultures of mouse and chicken cells.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2...8 °C