Eppendorf® 5430 Microcentrifuges
Supplier: Eppendorf
022620509 022620541 022620568 022620584 022620511 022620557 022620572 022620596 022620515 022620562 022620560 022620580 022620562 022620560 022620576
CA80094-172EA 9712.85 CAD
CA80094-172 CA80094-176 CA80094-178 CA80094-180 CA80094-182 CA97027-838 CA97027-840 CA97027-842 CA97027-844 CA97027-846 CA97027-850 89422-674 10030-956 10030-996 490015-774 CA10030-956 CA10030-996 75875-084 76460-822 76460-824
Eppendorf® 5430 Microcentrifuges
Centrifuges Microcentrifuges
The cross-over Eppendorf Centrifuge 5430 combines the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, it can accommodate rotors for microplates and 15/50 ml conical tubes as well as Eppendorf tubes and PCR strips.
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined
More Product Information