Chloride 2 Indicator Powder PIllows, Hach
Supplier: Hach
104399
CA97026-714PK 85.97 CAD
CA97026-714
Chloride 2 Indicator Powder PIllows, Hach
Environmental Test Kits
For determination of chloride by argentometric titration method with silver nitrate for test kits.
Ordering information: Pack of 100 Powder Pillows. Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.