EDTA 0.0716 M, cartridge for digital titrator

Supplier: Hach
Warning

Synonyms: Ethylenediaminetetraacetic acid tetrasodium salt, Ethylenedinitrilotetra-acetic acid tetrasodium salt, Tetrasodium ethylenediaminetetraacetate, EDTA tetrasodium, Edetic acid tetrasodium salt, Kestranal® 4S

2081701
CA97026-548EA 62.2 CAD
CA97026-548
EDTA 0.0716 M, cartridge for digital titrator
EDTA tetrasodium salt

EDTA standard solution


  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


For the determination of iron by Digital Titrator titration. 0.0716 M. Digits x 0.1 = mg/L Fe in 50 mL.

Formula: C₁₀H₁₂N₂Na₄O₈
MW: 380.17 g/mol
MDL Number: MFCD00150027
CAS Number: 64-02-8
Merck Index: 12,03557
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3da8dc42f6b4faeb40c2c5e98f983e37d310fe21&vid=c925ac956246bbfd17c182e83b57c15a3b8edfa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Method Name EDTA
Number of tests 40 - 100
Parameter EDTA
Platform Digital Titrator
Range 10 - 100 mg/L Fe
Titrant 0.0716 M EDTA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR