PathoScreen Medium Presence/Absence Powder Pillows, Hach
Supplier: Hach
2610696
CA97026-532PK 190.34 CAD
CA97026-532
PathoScreen Medium Presence/Absence Powder Pillows, Hach
Microbiology Test Systems Microorganism Tests
PathoScreen Medium Presence/Absence Powder Pillows, 100-mL sample.
  • Use to detect waterborne pathogens associated with fecal contamination of water sources
  • Simple presenceabsence method eliminates need for an incubator in tropical and semitropical climates
  • Complete Solutions for Testing Drinking Water
Ordering information: 50/pk.