Bag, Sterile, Whirl-Pak with Dechlorinating Agent, 177 mL, Hach
Supplier: Hach
2075333
CA97026-210PK 209.62 CAD
CA97026-210
Bag, Sterile, Whirl-Pak with Dechlorinating Agent, 177 mL, Hach
Bags Sample Bags
Specially designed for chlorinated water samples (Complies with USEPA and Standard Methods criteria).
  • Complete solutions for testing recreational water
  • Complete solutions for testing wastewater
  • Complete solutions for testing drinking water
Sterile (certified), disposable, transparent polyethylene bags with trusted 'Whirl-Pak' closure. Contains non-nutritive pill with 10 mg sodium thiosulfate (will dechlorinate up to 10 mg/Cl₂ in 100 ml sample ). Four ounce and 100 ml fill-lines. 'Write-on' strip accepts waterproof ink.
Ordering information: 100 per pack.