Conductivity Standard, 1990 µS/cm; (995 mg/L TDS, 1000 mg/L NaCl), 100 ml, Hach
Supplier: Hach
210542
CA97021-504EA 63.33 CAD
CA97021-504
Conductivity Standard, 1990 µS/cm; (995 mg/L TDS, 1000 mg/L NaCl), 100 ml, Hach
Standards Conductivity Standards
Sodium chloride standard solution
For calibrations and accuracy checks in conductivity measurements.