Chromolith® CapRod® Capillary Columns, MilliporeSigma
Supplier: MilliporeSigma
1.50402.0001 1.50403.0001 1.50405.0001 1.50407.0001 1.50409.0001 1.50400.0001 1.50403.0001 1.50405.0001 1.50407.0001 1.50409.0001 1.50400.0001 1.50402.0001 1.52031.0001 1.50402.0001 1.52031.0001 1.50404.0001 1.50424.0001 1.50426.0001
48219-914EA 2607.65 CAD
48219-914 97015-674 97015-678 97015-680 97015-682 97015-684 CA97015-674 CA97015-678 CA97015-680 CA97015-682 CA97015-684 EM1.50402.0001 EM1.52031.0001 CA1.50402.0001 CA1.52031.0001 CA1.50404.0001 CA1.50424.0001 CA1.50426.0001
Chromolith® CapRod® Capillary Columns, MilliporeSigma
Chromatography Columns
All Chromolith columns are made of monolithic silica with a characteristic bimodal pore structure.
  • Typical flow rates are above 1 µl/min
Chromolith CapRod is a capillary column that combines the speed of monolithic silica technology with the sensitivity of nano-LC, hence enabling new productivity levels for high-throughput proteomics LC applications. In contrast to microparticulate columns Chromolith CapRod can be operated at significantly higher flow rates without loss of performance. Separations can be achieved at 1 to 3 µl/min compared to 200 to 400More Product Information