Axial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0771116
CA97013-474EA 247.99 CAD
CA97013-474
Axial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Optical Windows
This axial view purge extension window is used with Optima™ 3x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments.
  • The window is made from high-grade quartz
  • Window should be replaced on a regular basis as part of a preventative maintenance schedule
  • Maintain factory performance using genuine PerkinElmer replacement parts
The Axial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics of the instrument.