Pocket Colorimeter II Test Kits
Supplier: Hach
2105528 2668000 2430000 2106069 2105569 2802246 2105669 1406499 2508025 2502025 2503025 2770900 2507025 2506025 2105628 1407099 2106169 2242000 2527025
CA21055-28PK 592.26 CAD
CA21055-28 CA2668000 CA58700-02 CA97009-434 CA97009-436 CA97009-438 CA97009-454 CA97009-456 CA97009-464 CA97009-470 CA97009-490 CA97009-494 CA97009-498 CA97009-500 CA97009-504 CA97009-512 CA97009-520 CA97009-536 CA97009-554 CA97009-560 CA97009-566 CA97009-468 CA97009-474 CA97009-488 CA97009-514
Pocket Colorimeter II Test Kits
Environmental Test Kits
This colorimeter is a portable, rugged, and convenient instrument, ideally suited for in-the-field analysis.
Tough, impact-resistant shell is also waterproof and floats. Four buttons allow easy operation. Accuracy is comparable to expensive lab instruments. All units are factory-programmed; no manual calibration is ever required. All units come with enough reagent sets for 100 tests.