OIML Class 1 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler Toledo
11123464 11123465 11123470 11123471 11123473 11123481 11123447 11123448 11123450 11123451 11123453 11123454 11123455 11123457 11123459 11123460 11123461 11123462 11123463 11123466 11123469 11123472 11123474 11123476 11123477 11123479 11123480 11123482 11123483 11123484 11123485 11123486 11123488 11123446 11123449 11123452 11123458 11123456 11123475 11123478 11123489 11123490 11123487
CA97007-438EA 1055 CAD
CA97007-438 CA97007-440 CA97007-446 CA97007-448 CA97007-452 CA97007-468 97007-404 97007-406 97007-410 97007-412 97007-416 97007-418 97007-420 97007-424 97007-428 97007-430 97007-432 97007-434 97007-436 97007-442 97007-444 97007-450 97007-454 97007-458 97007-460 97007-464 97007-466 97007-470 97007-472 97007-474 97007-476 97007-478 97007-482 97007-402 97007-408 97007-414 97007-426 97007-422 97007-456 CA97007-462 CA97007-484 89422-918 CA97007-480
OIML Class 1 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.