Super Erecta Shelf* Solid Shelving, Standard Stainless Steel, Metro*
Supplier: Metropolitan Wire
1424LS 1430LS 1830LS 1436FS 1442FS 1448FS 1824FS 1830FS 1842FS 1848FS 2124FS 2136FS 2142FS 2148FS 2160FS 2424FS 2430FS 1436LS 1442LS 1448LS 1460LS 1824LS 1836LS 1842LS 1860LS 2124LS 2130LS 2136LS 2142LS 2148LS 2160LS 2424LS 2430LS 2436LS 2442LS 1848LS 2448LS 1424FS 1430FS 1460FS 1836FS 1860FS 2130FS 2436FS 2442FS 2448FS 2460FS
CA97005-280EA 930.15 CAD
CA97005-280 CA97005-282 CA97005-284 CA97005-628 CA97005-630 CA97005-632 CA97005-634 CA97005-636 CA97005-638 CA97005-640 CA97005-642 CA97005-644 CA97005-646 CA97005-648 CA97005-650 CA97005-652 CA97005-654 CA97005-704 CA97005-706 CA97005-708 CA97005-710 CA97005-712 CA97005-714 CA97005-716 CA97005-718 CA97005-720 CA97005-722 CA97005-724 CA97005-726 CA97005-728 CA97005-730 CA97005-732 CA97005-734 CA97005-736 CA97005-738 CA97005-740 CA97005-800 CA97005-802 CA97005-822 CA97005-824 CA97005-826 CA97005-828 CA97005-830 CA97005-832 CA97005-834 CA97005-836 CA97005-838 CA97005-840
Super Erecta Shelf* Solid Shelving, Standard Stainless Steel, Metro*
Furniture Shelves
Solid shelving features 18-gauge stainless steel construction with flat, embossed, or louvered/embossed surfaces and black powder-coated corners.
  • Ship's edges help contain spillage minimize contamination and permit easier cleanup
  • Adjustable feet (bolt levelers) compensate for surface irregularities
Two-fold thickness forms a 3.2 mm (¹/₈") raised ship's edge on all four sides for all-around rigidity and high strength. Embossed or louvered/embossed shelf designs allow air circulation under packages and containers. Shelves can be loaded and unloaded fromMore Product Information